Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibenzocyclooctyne-Peg4-N-Hydroxysuccinimidyl Ester

Catalog No. 7640195MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-401-95MG 5 g
76-401-91MG 1 mg
76-401-950MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 76-401-95MG Supplier Sigma Aldrich Supplier No. 7640195MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Succinimidyl ester (NHS, amine reactive) functionalized cyclooctyne derivative for incorporation of the cyclooctyne moiety into amine containing compounds or biomolecules. Cyclooctynes are useful in strain-promoted copper-free click chemistry cycloaddition reactions. This dibenzocyclooctyne will react with azide functionalized compounds or biomolecules without the need for a Cu(I) catalyst to result in a stable triazole linkage. Applications Include: Protein-peptide conjugatesAntibody-enzyme or antibody-drug conjugatesProtein or peptide-oligonucleotide conjugates Surface modification

Specifications

CAS 1445086-12-3
Melting Point 219°C to 224°C
Molecular Formula C31H46NO3PPdS
Linear Formula C31H46NO3PPdS
Quantity 5 g
Synonym Palladiu mg3-Tricyclohexylphosphine; [(Tricyclohexylphosphine)-2-(2'-aminobiphenyl)]palladium(II) methanesulfonate
Molecular Weight (g/mol) 650.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.