Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzofuran-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D45675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4567-5G 5 g
D45671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4567-5G Supplier TCI America Supplier No. D45675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 100124-06-9
Molecular Formula C12H9BO3
MDL Number MFCD00092336
InChI Key ZXHUJRZYLRVVNP-UHFFFAOYSA-N
PubChem CID 2734328

Specifications

Melting Point 258°C
Color White
Quantity 5 g
Synonym dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid
SMILES OB(O)C1=CC=CC2=C1OC1=C2C=CC=C1
IUPAC Name {8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-6-yl}boronic acid
Molecular Weight (g/mol) 212.01
Formula Weight 212.01
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.