Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzofuran 98%, Thermo Scientific™

Catalog No. AC159300500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC159300500 50 g
AC159302500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159300500 Supplier Thermo Scientific Chemicals Supplier No. 159300500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-64-9
Molecular Formula C12H8O
Molecular Weight (g/mol) 168.195
MDL Number MFCD00004968
InChI Key TXCDCPKCNAJMEE-UHFFFAOYSA-N
Synonym dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide
PubChem CID 568
ChEBI CHEBI:28145
IUPAC Name dibenzofuran
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3O2

Specifications

Melting Point 81°C to 85°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 50 g
Beilstein 17, 70
Merck Index 15, 3013
Formula Weight 168.19
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-071-3
RTECSNumber HP4430000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.