Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzosuberol | 1210-34-0 | MFCD00003593 | >=98.0% | 210.27 | C15H14O | 10 G
SDP

Catalog No. 5000210066
Encompass_Preferred

Dibenzosuberol is a tricyclic research intermediate commonly used in synthetic and medicinal chemistry applications. It is provided as a solid research reagent with analytical documentation for laboratory use.

  • High purity: >=98.0%.
  • Solid, off-white to light yellow.
  • CAS number: 1210-34-0.
  • Molecular formula: C15H14O; molecular weight: 210.27 g/mol.
  • Available in multiple pack sizes, including 10 G.
  • Analytical documentation: data sheet and certificate of analysis available.

Catalog No. 50-002-10066 Supplier Medchemexpress LLC Supplier No. HYW04367610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dibenzosuberol is a tricyclic research intermediate commonly used in synthetic and medicinal chemistry applications. It is provided as a solid research reagent with analytical documentation for laboratory use.

  • High purity: >=98.0%.
  • Solid, off-white to light yellow.
  • CAS number: 1210-34-0.
  • Molecular formula: C15H14O; molecular weight: 210.27 g/mol.
  • Available in multiple pack sizes, including 10 G.
  • Analytical documentation: data sheet and certificate of analysis available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.