Learn More
Medchemexpress LLC Dibenzosuberone | 1210-35-1 | 99.8% | C15H12O | 100 G
SDP
Catalog No. 5000434846
Encompass_Preferred
Dibenzosuberone is part of the tricyclic antidepressant drug family (TCAs) and is utilized in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia. It also serves as a crucial intermediate for synthesizing various active pharmaceutical ingredients (APIs).
  • Part of tricyclic antidepressant drug family (TCAs).
  • Utilized in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia.
  • Serves as a crucial intermediate for synthesizing various active pharmaceutical ingredients (APIs) such as Amitriptyline, Nortriptyline, Noxiptiline, and Cyproheptadine.
Catalog No. 50-004-34846 Supplier Medchemexpress LLC Supplier No. HY78509100G
Dibenzosuberone is part of the tricyclic antidepressant drug family (TCAs) and is utilized in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia. It also serves as a crucial intermediate for synthesizing various active pharmaceutical ingredients (APIs).
  • Part of tricyclic antidepressant drug family (TCAs).
  • Utilized in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia.
  • Serves as a crucial intermediate for synthesizing various active pharmaceutical ingredients (APIs) such as Amitriptyline, Nortriptyline, Noxiptiline, and Cyproheptadine.