Learn More
Medchemexpress LLC Dibenzosuberone | 1210-35-1 | 99.8% | C15H12O | 50 G
SDP
Catalog No. 5000433291
Encompass_Preferred
Dibenzosuberone belongs to the family of tricyclic antidepressant drugs (TCAs). It is commonly used in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia. It also serves as a key intermediate in the synthesis of various active pharmaceutical ingredients (APIs).
  • Belongs to the family of tricyclic antidepressant drugs (TCAs).
  • Used in research on migraine, tension headaches, anxiety disorders, and schizophrenia.
  • Key intermediate in the synthesis of various active pharmaceutical ingredients (APIs).
Catalog No. 50-004-33291 Supplier Medchemexpress LLC Supplier No. HY7850950G
Dibenzosuberone belongs to the family of tricyclic antidepressant drugs (TCAs). It is commonly used in research concerning migraine, tension headaches, anxiety disorders, and schizophrenia. It also serves as a key intermediate in the synthesis of various active pharmaceutical ingredients (APIs).
  • Belongs to the family of tricyclic antidepressant drugs (TCAs).
  • Used in research on migraine, tension headaches, anxiety disorders, and schizophrenia.
  • Key intermediate in the synthesis of various active pharmaceutical ingredients (APIs).