Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzosuberone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Density:
1.156 g/mL (at 25°C (literature))
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Density
11-102-2949 1.156 g/mL (at 25°C (literature))
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2949 Supplier MilliporeSigma Supelco Supplier No. BP858
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1210-35-1
Melting Point 32°C to 34°C (literature)
Density 1.156 g/mL (at 25°C (literature))
Boiling Point 148°C/0.3 mmHg (literature)
Flash Point 113°C (Closed Cup)
Molecular Formula C15H12O
Refractive Index n20/D 1.6332 (literature)
Linear Formula C15H12O
Quantity 1 Ea.
MDL Number MFCD00003587
Synonym 5-Dibenzosuberone; 10,11-Dihydro-5 H-dibenzo[a,d]cyclohepten-5-one; Dibenzocycloheptadienone
Formula Weight 208.26
Grade BP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.