Learn More
Medchemexpress LLC Dibenzothiophene | 132-65-0 | MFCD00004969 | 99.9% | 25 G
SDP
Catalog No. 5000405644
Encompass_Preferred
Dibenzothiophene is an orally active and noncompetitive CYP1A inhibitor, primarily used for studying developmental toxicity mechanisms. It inhibits CYP1A-mediated EROD activity with a Km of 0.592 μM and interacts with the AHR pathway. This compound enhances the embryotoxicity of β-naphthoflavone and shows acute toxicity in mice.
  • Orally active and noncompetitive CYP1A inhibitor
  • Inhibits CYP1A-mediated EROD activity
  • Interacts with the AHR pathway
  • Enhances embryotoxicity of β-naphthoflavone
  • Used for studying mechanisms of developmental toxicity
Catalog No. 50-004-05644 Supplier Medchemexpress LLC Supplier No. 25GHYB0973
Dibenzothiophene is an orally active and noncompetitive CYP1A inhibitor, primarily used for studying developmental toxicity mechanisms. It inhibits CYP1A-mediated EROD activity with a Km of 0.592 μM and interacts with the AHR pathway. This compound enhances the embryotoxicity of β-naphthoflavone and shows acute toxicity in mice.
  • Orally active and noncompetitive CYP1A inhibitor
  • Inhibits CYP1A-mediated EROD activity
  • Interacts with the AHR pathway
  • Enhances embryotoxicity of β-naphthoflavone
  • Used for studying mechanisms of developmental toxicity