Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D43735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4373-5G 5 g
D43731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4373-5G Supplier TCI America Supplier No. D43735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 668983-97-9
Molecular Formula C12H9BO2S
MDL Number MFCD01318982
InChI Key CSLSCVHILGCSTE-UHFFFAOYSA-N
PubChem CID 2794660

Specifications

Color White
Quantity 5 g
Synonym dibenzo b,d thiophen-2-ylboronic acid, dibenzothiophene-2-boronic acid, dibenzo b,d thien-2-ylboronic acid, dibenzothiophen-2-yl-dihydroxy-borane, boronic acid, 2-dibenzothienyl
SMILES OB(O)C1=CC2=C(SC3=C2C=CC=C3)C=C1
IUPAC Name {8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaen-4-yl}boronic acid
Molecular Weight (g/mol) 228.07
Formula Weight 228.07
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.