Learn More
Medchemexpress LLC Dibenzothiophene-d8 | 33262-29-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0973S5MG
Encompass_Preferred
Dibenzothiophene-d8 is a deuterium-labeled version of Dibenzothiophene, a noncompetitive CYP1A inhibitor that interacts with the AHR pathway. It is primarily used to study the mechanism of developmental toxicity. Dibenzothiophene-d8 can serve as a tracer or internal standard for quantitative analysis.
  • Deuterium-labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS
  • Useful for studying developmental toxicity mechanisms
  • Deuteration potentially affects pharmacokinetic and metabolic profiles
Catalog No. 50-004-07860