Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzothiophene, MilliporeSigma™ Supelco™

Catalog No. 111020065 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0065 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0065 Supplier MilliporeSigma Supelco Supplier No. 45776250MG
Add to Cart
Add to Cart
Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 132-65-0
Physical Form Neat
Linear Formula C12H8 S
MDL Number MFCD00004969
Melting Point 97°C to 100°C
Boiling Point 332°C to 333°C
Synonym DBT; Diphenylene sulfide
Type Analytical Standard
Quantity 250 mg
Formula Weight 184.26
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.