Learn More
Medchemexpress LLC (+)-Dibenzoyl-D-tartaric acid | 17026-42-5 | 63222 | 99.8% | 358.30 | 100 G
SDP
Catalog No. 5000401208
Encompass_Preferred
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent used as a biological material or organic compound for life science-related research. It serves as a reagent in the preparation of chiral salts and for the chiral resolution of amino compounds.
  • Used as a biochemical reagent in life science research.
  • Functions as a reagent for preparing chiral salts.
  • Aids in the chiral resolution of amino compounds.
Catalog No. 50-004-01208 Supplier Medchemexpress LLC Supplier No. HY20551100G
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent used as a biological material or organic compound for life science-related research. It serves as a reagent in the preparation of chiral salts and for the chiral resolution of amino compounds.
  • Used as a biochemical reagent in life science research.
  • Functions as a reagent for preparing chiral salts.
  • Aids in the chiral resolution of amino compounds.