Learn More
Medchemexpress LLC (+)-Dibenzoyl-D-tartaric acid | 17026-42-5 | 99.8% | 358.30 | 25 G
SDP
Catalog No. 5000402858
Encompass_Preferred
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent suitable for life science research. It can be used as a biological material or organic compound.
  • Used as a reagent in the preparation of chiral salts.
  • Serves as a reagent for the chiral resolution of amino compounds.
Catalog No. 50-004-02858 Supplier Medchemexpress LLC Supplier No. 25GHY20551
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent suitable for life science research. It can be used as a biological material or organic compound.
  • Used as a reagent in the preparation of chiral salts.
  • Serves as a reagent for the chiral resolution of amino compounds.