Learn More
Medchemexpress LLC (+)-Dibenzoyl-D-tartaric acid | 17026-42-5 | 99.9% | 358.30 | 1 ML
SDP
Catalog No. 5000401891
Encompass_Preferred
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. For chiral resolution, it is used as a reagent in the preparation of chiral salts and also serves as a reagent for chiral resolution of amino compounds.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used as a reagent in the preparation of chiral salts
  • Serves as a reagent for chiral resolution of amino compounds
Catalog No. 50-004-01891 Supplier Medchemexpress LLC Supplier No. HY2055110MM/1ML
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. For chiral resolution, it is used as a reagent in the preparation of chiral salts and also serves as a reagent for chiral resolution of amino compounds.
  • Biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Used as a reagent in the preparation of chiral salts
  • Serves as a reagent for chiral resolution of amino compounds