Learn More
Medchemexpress LLC (+)-Dibenzoyl-D-tartaric acid | 17026-42-5 | 99.9% | 358.30 | 1000 G
SDP
Catalog No. 5000401055
Encompass_Preferred
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent. It can be used as a biological material or organic compound for life science related research.
  • Reagent for chiral resolution
  • Used in the preparation of chiral salts
  • Serves as a reagent for chiral resolution of amino compounds
Catalog No. 50-004-01055 Supplier Medchemexpress LLC Supplier No. HY205511000G
(+)-Dibenzoyl-D-tartaric acid is a biochemical reagent. It can be used as a biological material or organic compound for life science related research.
  • Reagent for chiral resolution
  • Used in the preparation of chiral salts
  • Serves as a reagent for chiral resolution of amino compounds