Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dibenzoyl-D-tartaric acid, 98+%

Catalog No. AC302515000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302515000 500 g Plastic bottle
AC302510250 25 g Plastic bottle
AC302511000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC302515000 Supplier Thermo Scientific Chemicals Supplier No. 302515000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17026-42-5 | C18H14O8 | 358.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Specific Rotation Condition +120° (20°C c=5,CH3OH)
Specific Rotation + 120
Solubility Information Solubility in water: insoluble
IUPAC Name (2S,3S)-2,3-dibenzoyloxybutanedioic acid
Formula Weight 358.31
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-097-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.