Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Dibenzoyl-L-tartaric anhydride, 98%

Catalog No. AC336030250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC336030250 25 g Plastic bottle
AC336030050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC336030250 Supplier Thermo Scientific Chemicals Supplier No. 336030250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64339-95-3 | C18H12O7 | 340.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64339-95-3
Molecular Formula C18H12O7
Molecular Weight (g/mol) 340.29
MDL Number MFCD00067102
InChI Key OXIKRMSPXYQFOT-ZCWZLOQUNA-N
Synonym 3r,4r-2,5-dioxotetrahydrofuran-3,4-diyl dibenzoate, +-dibenzoyl-l-tartaric anhydride, 2,5-furandione, 3,4-bis benzoyloxy dihydro-, 3r,4r, 2-o,3-o-dibenzoyl-l-tartaric anhydride, 3r,4r-2,5-dioxo-3,4-dibenzoyloxy-3,4-dihydrofuran, 3r,4r-4-benzoyloxy-2,5-dioxooxolan-3-yl benzoate
PubChem CID 2733743
IUPAC Name [(3R,4R)-4-benzoyloxy-2,5-dioxooxolan-3-yl] benzoate
SMILES O=C(O[C@@H]1[C@@H](OC(=O)C2=CC=CC=C2)C(=O)OC1=O)C1=CC=CC=C1

Specifications

Melting Point 174°C
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethyl ether
Formula Weight 340.29
Percent Purity 98%
Chemical Name or Material (+)-Dibenzoyl-L-tartaric anhydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.