Learn More
Medchemexpress LLC Dibenzoyl thiamine | 299-88-7 | 99.8% | 50 MG
SDP
Catalog No. 5000412290
Encompass_Preferred
Dibenzoyl Thiamine (Bentiamine) is a derivative of thiamine that is rapidly absorbed into the body and converted to thiamine. It is a thiol-type thiamine composed of thiamine and benzoic acid. It is not decomposed by aneurinase, making it more suitable for food processing than thiamine hydrochloride. This product is for research use only.
  • Rapidly absorbed and converted to thiamine
  • Composed of thiamine and benzoic acid
  • Not decomposed by aneurinase
  • Suitable for food processing
  • Non-toxic and non-mutagenic in studies
Catalog No. 50-004-12290 Supplier Medchemexpress LLC Supplier No. HYB221250MG
Dibenzoyl Thiamine (Bentiamine) is a derivative of thiamine that is rapidly absorbed into the body and converted to thiamine. It is a thiol-type thiamine composed of thiamine and benzoic acid. It is not decomposed by aneurinase, making it more suitable for food processing than thiamine hydrochloride. This product is for research use only.
  • Rapidly absorbed and converted to thiamine
  • Composed of thiamine and benzoic acid
  • Not decomposed by aneurinase
  • Suitable for food processing
  • Non-toxic and non-mutagenic in studies