Learn More
Medchemexpress LLC Dibenzoyl Thiamine (Bentiamine) | 299-88-7 | 99.8% | 1 ML
SDP
Catalog No. 5000410297
Encompass_Preferred
Dibenzoyl Thiamine (Bentiamine) is a thiamine derivative that is rapidly absorbed and converted to thiamine. This thiol-type thiamine is composed of thiamine and benzoic acid, and is not decomposed by aneurinase (thiamine-decomposing enzyme), making it more suitable for food processing than thiamine hydrochloride.
  • Thiol-type thiamine derivative.
  • Not decomposed by aneurinase.
  • Rapidly absorbed into the body.
  • Converted to thiamine (Vitamin B1).
  • Suitable for food processing.
  • Metabolized to Vitamin B1 and benzoic acid in vivo.
  • No specific toxic, embryotoxic, teratogenic, or mutagenic effects observed in studies.
  • Solid appearance, white to off-white color.
Catalog No. 50-004-10297 Supplier Medchemexpress LLC Supplier No. HYB221210MM/1ML
Dibenzoyl Thiamine (Bentiamine) is a thiamine derivative that is rapidly absorbed and converted to thiamine. This thiol-type thiamine is composed of thiamine and benzoic acid, and is not decomposed by aneurinase (thiamine-decomposing enzyme), making it more suitable for food processing than thiamine hydrochloride.
  • Thiol-type thiamine derivative.
  • Not decomposed by aneurinase.
  • Rapidly absorbed into the body.
  • Converted to thiamine (Vitamin B1).
  • Suitable for food processing.
  • Metabolized to Vitamin B1 and benzoic acid in vivo.
  • No specific toxic, embryotoxic, teratogenic, or mutagenic effects observed in studies.
  • Solid appearance, white to off-white color.