Learn More
Medchemexpress LLC Dibenzoylmethane | 120-46-7 | 98.0% | 224.26 | 10 G
SDP
Catalog No. 5000453382
Encompass_Preferred
Dibenzoylmethane is an analytical standard that functions as a Keap1-Nrf2 activator. A minor ingredient found in licorice, it is known to activate Nrf2, preventing various cancers and oxidative damage. As an analog of curcumin, Dibenzoylmethane facilitates the dissociation from Keap1 and promotes the nuclear translocation of Nrf2. It is provided as an off-white to light yellow solid and is intended for research and analytical applications.
  • Activates Nrf2
  • Prevents various cancers and oxidative damage
  • Analog of curcumin
  • Results in dissociation from Keap1
  • Induces nuclear translocation of Nrf2
  • Off-white to light yellow solid
  • Intended for research and analytical applications
Catalog No. 50-004-53382 Supplier Medchemexpress LLC Supplier No. HYW00973110G
Dibenzoylmethane is an analytical standard that functions as a Keap1-Nrf2 activator. A minor ingredient found in licorice, it is known to activate Nrf2, preventing various cancers and oxidative damage. As an analog of curcumin, Dibenzoylmethane facilitates the dissociation from Keap1 and promotes the nuclear translocation of Nrf2. It is provided as an off-white to light yellow solid and is intended for research and analytical applications.
  • Activates Nrf2
  • Prevents various cancers and oxidative damage
  • Analog of curcumin
  • Results in dissociation from Keap1
  • Induces nuclear translocation of Nrf2
  • Off-white to light yellow solid
  • Intended for research and analytical applications