Learn More
Medchemexpress LLC Dibenzoylmethane | 120-46-7 | 98.0% | 224.26 | 100 G
SDP
Catalog No. 5000453330
Encompass_Preferred
Dibenzoylmethane is a minor ingredient found in licorice that activates Nrf2. It helps prevent various cancers and oxidative damage. As an analog of curcumin, it causes the dissociation from Keap1 and nuclear translocation of Nrf2.
  • Activates Nrf2
  • Helps prevent various cancers
  • Helps prevent oxidative damage
  • Analog of curcumin
  • Causes dissociation from Keap1 and nuclear translocation of Nrf2
Catalog No. 50-004-53330 Supplier Medchemexpress LLC Supplier No. HYW009731100G
Dibenzoylmethane is a minor ingredient found in licorice that activates Nrf2. It helps prevent various cancers and oxidative damage. As an analog of curcumin, it causes the dissociation from Keap1 and nuclear translocation of Nrf2.
  • Activates Nrf2
  • Helps prevent various cancers
  • Helps prevent oxidative damage
  • Analog of curcumin
  • Causes dissociation from Keap1 and nuclear translocation of Nrf2