Learn More
Medchemexpress LLC Dibenzoylmethane | 120-46-7 | MFCD00003085 | 98.0% | 224.26 | 5 G
SDP
Catalog No. 5000452068
Encompass_Preferred
Dibenzoylmethane, a minor ingredient in licorice, activates Nrf2 and prevents various cancers and oxidative damage. It is an analog of curcumin that causes dissociation from Keap1 and nuclear translocation of Nrf2. This product is for research use only and not intended for sale to patients.
  • Activates Nrf2
  • Prevents various cancers and oxidative damage
  • Analog of curcumin
  • Causes dissociation from Keap1 and nuclear translocation of Nrf2
Catalog No. 50-004-52068 Supplier Medchemexpress LLC Supplier No. HYW0097315G
Dibenzoylmethane, a minor ingredient in licorice, activates Nrf2 and prevents various cancers and oxidative damage. It is an analog of curcumin that causes dissociation from Keap1 and nuclear translocation of Nrf2. This product is for research use only and not intended for sale to patients.
  • Activates Nrf2
  • Prevents various cancers and oxidative damage
  • Analog of curcumin
  • Causes dissociation from Keap1 and nuclear translocation of Nrf2