Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzyl disulfide | 150-60-7 | 99.9% | 246.39 | 1 ML
SDP

Catalog No. 5000451676
Encompass_Preferred

Dibenzyl disulfide is an endogenous metabolite for research use.

  • Purity of 99.94%
  • Molecular weight of 246.39
  • Chemical formula C14H14S2
  • CAS number 150-60-7
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage for solutions at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-51676 Supplier Medchemexpress LLC Supplier No. HYW00951610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzyl disulfide is an endogenous metabolite for research use.

  • Purity of 99.94%
  • Molecular weight of 246.39
  • Chemical formula C14H14S2
  • CAS number 150-60-7
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder at -20°C for 3 years or 4°C for 2 years
  • Recommended storage for solutions at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.