Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzyl disulfide | 150-60-7 | MFCD00004783 | 99.9% | 246.39 | 500 G
SDP

Catalog No. 5000366908
Encompass_Preferred

Dibenzyl disulfide is an endogenous metabolite. It is a solid with a white to off-white appearance.

  • Purity: 99.94%
  • Molecular Weight: 246.39
  • CAS number: 150-60-7
  • Formula: C14H14S2
  • Initial source: Plants Petiveriaceae Petiveria alliacea
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-66908 Supplier Medchemexpress LLC Supplier No. HYW009516500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzyl disulfide is an endogenous metabolite. It is a solid with a white to off-white appearance.

  • Purity: 99.94%
  • Molecular Weight: 246.39
  • CAS number: 150-60-7
  • Formula: C14H14S2
  • Initial source: Plants Petiveriaceae Petiveria alliacea
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.