Learn More
Apexbio Technology LLC DIBENZYL-DISULFIDE-1G
SDP
Catalog No. 5000568220
Encompass_Preferred
Dibenzyl disulfide (CAS No 150-60-7) is a significant compound in biomedical research particularly in the study of oxidative stress and cell signaling pathways Originating from natural sources like Allium vegetables it is known for its potential to modulate biochemical pathways related to enzymatic antioxidant activities Research indicates its influence on the reduction of oxidative stress via the regulation of reactive oxygen species Dibenzyl disulfide exhibits common in vitro activity in the nanomolar to low micromolar range It is utilized in various applications including cell models examining cancer cell lines and animal studies exploring its pharmacological effects Additionally its role in drug screening highlights its potential as an adjunct or alternative therapeutic agent Experimental concentrations typically depend on the specific research design ensuring adaptability across different study frameworks and objectives
Catalog No. 50-005-68220 Supplier Apexbio Technology LLC Supplier No. C68241000
Dibenzyl disulfide (CAS No 150-60-7) is a significant compound in biomedical research particularly in the study of oxidative stress and cell signaling pathways Originating from natural sources like Allium vegetables it is known for its potential to modulate biochemical pathways related to enzymatic antioxidant activities Research indicates its influence on the reduction of oxidative stress via the regulation of reactive oxygen species Dibenzyl disulfide exhibits common in vitro activity in the nanomolar to low micromolar range It is utilized in various applications including cell models examining cancer cell lines and animal studies exploring its pharmacological effects Additionally its role in drug screening highlights its potential as an adjunct or alternative therapeutic agent Experimental concentrations typically depend on the specific research design ensuring adaptability across different study frameworks and objectives