Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzyl hydrazodicarboxylate | 5394-50-3 | C16H16N2O4 | 50 MG
SDP

Catalog No. 5000434116
Encompass_Preferred

Dibenzyl hydrazodicarboxylate is a protein cross-linking agent. This chemical is intended for research use only and is not for human use.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-34116 Supplier Medchemexpress LLC Supplier No. HY7621450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzyl hydrazodicarboxylate is a protein cross-linking agent. This chemical is intended for research use only and is not for human use.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.