Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibenzyl hydrazodicarboxylate | 5394-50-3 | MFCD00022028 | 300.31 | C16H16N2O4 | 10 MG
SDP

Catalog No. 5000298801
Encompass_Preferred

Dibenzyl hydrazodicarboxylate is a small-molecule protein cross-linking reagent supplied for research use. It is used to cross-link proteins and other biomolecules in biochemical and structural studies.

  • Protein cross-linking reagent for biochemical applications.
  • CAS number 5394-50-3; molecular formula C16H16N2O4; molecular weight 300.31.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.
  • Purity and appearance not specified by the manufacturer.

Catalog No. 50-002-98801 Supplier Medchemexpress LLC Supplier No. HY7621410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibenzyl hydrazodicarboxylate is a small-molecule protein cross-linking reagent supplied for research use. It is used to cross-link proteins and other biomolecules in biochemical and structural studies.

  • Protein cross-linking reagent for biochemical applications.
  • CAS number 5394-50-3; molecular formula C16H16N2O4; molecular weight 300.31.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.
  • Purity and appearance not specified by the manufacturer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.