Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzyl phosphite, 90+%, technical

Catalog No. AC112661000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112661000 100 g Glass bottle
AC112660050 5 g Glass bottle
AC112660250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC112661000 Supplier Thermo Scientific Chemicals Supplier No. 112661000
  • $385.00
    $327.65 / Each of 1

    Save $57.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17176-77-1 | C14H14O3P | 261.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17176-77-1
Molecular Formula C14H14O3P
Molecular Weight (g/mol) 261.24
MDL Number MFCD00004774
InChI Key RQKYHDHLEMEVDR-UHFFFAOYSA-N
Synonym dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
PubChem CID 6334615
SMILES O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.1800g/mL
Boiling Point 110.0°C to 120.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5560
Linear Formula (C6H5CH2O)2P(O)H
Quantity 100 g
Fieser 01,198
Specific Gravity 1.18
Merck Index 15,3019
IUPAC Name oxo-bis(phenylmethoxy)phosphanium
Formula Weight 262.23
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Dibenzyl phosphite, tech., 80%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-226-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.