Learn More
Cayman Chemical DIbenzylfluoresceIn 10mg
SDP
Catalog No. 502965157
Encompass_Preferred
A fluorogenic probe that acts as a substrate for specific cytochrome P450 (CYP) isoforms, including CYP3A4, CYP2C8, CYP2C9, CYP2C19, and aromatase (CYP19); used to detect changes in CYP catalytic activity caused by drugs or disease
Catalog No. 50-296-5157 Supplier Cayman Chemical Supplier No. 1680810 MG
A fluorogenic probe that acts as a substrate for specific cytochrome P450 (CYP) isoforms, including CYP3A4, CYP2C8, CYP2C9, CYP2C19, and aromatase (CYP19); used to detect changes in CYP catalytic activity caused by drugs or disease