Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibrom™, MilliporeSigma™ Supelco™

Catalog No. 111020466 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0466 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0466 Supplier MilliporeSigma Supelco Supplier No. 45429250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line Dibrom
CAS 300-76-5
Physical Form Neat
Linear Formula C4H7 Br2 Cl2O4 P
MDL Number MFCD00053227
Synonym (1,2-Dibromo-2,2-dichloroethyl) dimethyl phosphate; Bromchlophos; Naled
Type Analytical Standard
Quantity 250 mg
Formula Weight 380.78
Grade Analytical Standard

Dibrom is a registered trademark of AMVAC Chemical Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.