Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific™

Catalog No. AA4666903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4666903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4666903 Supplier Thermo Scientific Chemicals Supplier No. 4666903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000310-63-3
Molecular Formula C26H44Br2FeP2Pd
Molecular Weight (g/mol) 740.66
MDL Number MFCD14155687
InChI Key SSCAGSSZVQHQTF-UHFFFAOYSA-L
Synonym pdbr2 dtbpf, 1,1'-bis di-tert-butylphosphino ferrocene dibromopalladium ii, dibromo 1,1'-bis di-tert-butylphosphino ferrocene palladium ii
PubChem CID 73994980
IUPAC Name dibromopalladium;ditert-butyl(cyclopenta-2,4-dien-1-yl)phosphane;iron(2+)
SMILES [Fe].Br[Pd++]Br.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

Specifications

Color Brown
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in dichloromethane.
Formula Weight 740.66
Physical Form Crystalline Solid
Chemical Name or Material Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.