Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromo[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II), Thermo Scientific™

Catalog No. AA4666403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4666403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4666403 Supplier Thermo Scientific Chemicals Supplier No. 4666403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366488-99-5
Molecular Formula C44H34Br2P2Pd+2
Molecular Weight (g/mol) 890.932
MDL Number MFCD14155710
InChI Key XOOQZGNLBGHEEX-UHFFFAOYSA-N
Synonym dibromo 2,2'-bis diphenylphosphino-1,1'-binaphthyl palladium ii, pdbr2 binap, 2,2'-bis diphenylphosphino-1,1'-binaphthyl dibromopalladium ii
PubChem CID 73994988
IUPAC Name dibromopalladium;[1-(2-diphenylphosphaniumylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphanium
SMILES C1=CC=C(C=C1)[PH+](C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)[PH+](C7=CC=CC=C7)C8=CC=CC=C8.Br[Pd]Br

Specifications

Color Orange
Quantity 1 g
Formula Weight 888.91
Physical Form Crystalline Solid
Chemical Name or Material Dichloro[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II)
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.