Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromoacetonitrile, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018420 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8420 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8420 Supplier MilliporeSigma Supelco Supplier No. 442552
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibromoacetonitrile belongs to the family of dihaloacetonitrile compounds that are volatile chlorination by-products. They are proven to be carcinogenic and mutagenic to mice.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 3252-43-5
Density 2.296 g/mL (at 25°C)
Physical Form Neat
Linear Formula Br2 CHCN
MDL Number MFCD00001856
Boiling Point 67°C to 69°C (24 mmHg)
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 198.84
Percent Purity ≥90% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.