Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibromocarbazole | 136630-39-2 | 99.9% | 325.00 | 10 G
SDP

Catalog No. 5000404059
Encompass_Preferred

2,7-Dibromocarbazole is a biochemical reagent intended for life science research, functioning as a biological material or organic compound. This product is for laboratory use only, suitable for the manufacture of substances by qualified scientists in equipped facilities, and is not for sale to patients.

  • Functions as a biological material or organic compound
  • Suitable for life science research
  • Intended for laboratory and research use only
  • Identified for the manufacture of substances
  • Stable under recommended storage conditions
  • Targeted for biochemical assay reagents
  • Soluble in DMSO at 100 mg/mL
  • Appearance is a white to off-white solid
  • Product complies with given specifications
  • Not validated for medical applications

Catalog No. 50-004-04059 Supplier Medchemexpress LLC Supplier No. HY3446410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,7-Dibromocarbazole is a biochemical reagent intended for life science research, functioning as a biological material or organic compound. This product is for laboratory use only, suitable for the manufacture of substances by qualified scientists in equipped facilities, and is not for sale to patients.

  • Functions as a biological material or organic compound
  • Suitable for life science research
  • Intended for laboratory and research use only
  • Identified for the manufacture of substances
  • Stable under recommended storage conditions
  • Targeted for biochemical assay reagents
  • Soluble in DMSO at 100 mg/mL
  • Appearance is a white to off-white solid
  • Product complies with given specifications
  • Not validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.