Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibromoisocyanuric Acid 97.0+%, TCI America™
SDP

Catalog No. D375325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D375325G 25 g
D37535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D375325G Supplier TCI America Supplier No. D375325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15114-43-9
Molecular Formula C3HBr2N3O3
Molecular Weight (g/mol) 286.867
MDL Number MFCD00463941
InChI Key HHBCEKAWSILOOP-UHFFFAOYSA-N
Synonym 1,3-Dibromo-1,3,5-triazine-2,4,6-trione, DBI
PubChem CID 192912
IUPAC Name 1,3-dibromo-1,3,5-triazinane-2,4,6-trione
SMILES C1(=O)NC(=O)N(C(=O)N1Br)Br

Specifications

Melting Point 309°C
Quantity 25 g
UN Number 3085
Formula Weight 286.87
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibromoisocyanuric Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.