Learn More
Medchemexpress LLC Dibromomaleimide-C5-COOH | 1443214-97-8 | 99.4% | 369.01 | C10H11Br2NO4 | 5 MG
SDP
Catalog No. 5000204320
Encompass_Preferred
Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide (DBM) linker used in antibody-drug conjugate (ADC) synthesis. It provides a C5 carboxylic acid handle for conjugation to payloads such as MMAF and is supplied for research use only.
  • Bifunctional dibromomaleimide linker for ADC synthesis.
  • Suitable for conjugation to MMAF and similar payloads.
  • High purity (99.4%) to support reproducible chemistry.
  • Molecular weight 369.01; formula C10H11Br2NO4.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-04320 Supplier Medchemexpress LLC Supplier No. HY1570805MG
Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide (DBM) linker used in antibody-drug conjugate (ADC) synthesis. It provides a C5 carboxylic acid handle for conjugation to payloads such as MMAF and is supplied for research use only.
  • Bifunctional dibromomaleimide linker for ADC synthesis.
  • Suitable for conjugation to MMAF and similar payloads.
  • High purity (99.4%) to support reproducible chemistry.
  • Molecular weight 369.01; formula C10H11Br2NO4.
  • Intended for research use only; not for human or clinical use.