Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibromomaleimide-C5-COOH | 1443214-97-8 | 99.4% | 369.01 g/mol | C10H11Br2NO4 | 10 MG
SDP

Catalog No. 5000204368
Encompass_Preferred

Dibromomaleimide-C5-COOH (DBM-C5-COOH) is a bifunctional dibromomaleimide linker used in antibody-drug conjugate (ADC) chemistry to couple cytotoxic payloads such as MMAF. It is supplied as a high-purity small molecule reagent with good solubility in DMSO and defined storage recommendations.

  • Bifunctional dibromomaleimide linker suitable for conjugation chemistry.
  • High reported purity of 99.4% for reproducible results.
  • Soluble in DMSO at ≥100 mg/mL for convenient stock solutions.
  • Available in small research quantities for exploratory ADC development.
  • Storage recommendations provided to maintain stability.

Catalog No. 50-002-04368 Supplier Medchemexpress LLC Supplier No. HY15708010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dibromomaleimide-C5-COOH (DBM-C5-COOH) is a bifunctional dibromomaleimide linker used in antibody-drug conjugate (ADC) chemistry to couple cytotoxic payloads such as MMAF. It is supplied as a high-purity small molecule reagent with good solubility in DMSO and defined storage recommendations.

  • Bifunctional dibromomaleimide linker suitable for conjugation chemistry.
  • High reported purity of 99.4% for reproducible results.
  • Soluble in DMSO at ≥100 mg/mL for convenient stock solutions.
  • Available in small research quantities for exploratory ADC development.
  • Storage recommendations provided to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.