Learn More
Medchemexpress LLC Dibromomaleimide-C5-COOH (DBM-C5-COOH) | 1443214-97-8 | MFCD34705293 | 99.4% | 369.01 | C10H11Br2NO4 | 25 MG
SDP
Catalog No. 5000204466
Encompass_Preferred
Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide linker used for the construction of antibody-drug conjugates (ADCs), enabling attachment of payloads such as MMAF. It is supplied with high reported purity and is soluble in DMSO at high concentration.
  • Bifunctional dibromomaleimide linker for ADC assembly.
  • Can be used to connect MMAF to antibodies.
  • High reported purity, 99.4%.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Available in small milligram package sizes for research use.
Catalog No. 50-002-04466 Supplier Medchemexpress LLC Supplier No. HY15708025MG
Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide linker used for the construction of antibody-drug conjugates (ADCs), enabling attachment of payloads such as MMAF. It is supplied with high reported purity and is soluble in DMSO at high concentration.
  • Bifunctional dibromomaleimide linker for ADC assembly.
  • Can be used to connect MMAF to antibodies.
  • High reported purity, 99.4%.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Available in small milligram package sizes for research use.