Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibromomethane

Catalog No. D41686500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41686500ML 500 mL
D416862L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41686500ML Supplier Sigma Aldrich Supplier No. D41686500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibromomethane

Specifications

CAS 74-95-3
Melting Point -52°C (lit.)
Density 2.477 g/mL (at 25°C (literature))
Boiling Point 96°C to 98°C (lit.)
Molecular Formula CH2Br2
Refractive Index n20/D 1.541 (literature)
Linear Formula CH2Br2
MDL Number MFCD00000168
Quantity 500 mL
Synonym Methylene bromide
Molecular Weight (g/mol) 173.83
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.