Learn More
AdipoGen Dibucaine hydrochloride
SDP
Catalog No. 502561348
Encompass_Preferred
Chemical. CAS 61-12-1. Formula C20H29N3O2 . HCl. MW 343.4 . 36.5. Synthetic. Dibucaine is a long-acting local anesthetic. Potent Na channel blocker. It blocks both the initiation and conduction of nerve impulses by decreasing the neuronal membranes permeability to sodium ions. This reversibly stabilizes the membrane and inhibits depolarization, resulting in the failure of a propagated action potential and subsequent conduction blockade. Induces apoptosis through calpain, cytochrome oxidase etc, and shows antiviral and antiparasitic activity.
Catalog No. 50-256-1348 Supplier AdipoGen Supplier No. CDXC0693G025
Chemical. CAS 61-12-1. Formula C20H29N3O2 . HCl. MW 343.4 . 36.5. Synthetic. Dibucaine is a long-acting local anesthetic. Potent Na channel blocker. It blocks both the initiation and conduction of nerve impulses by decreasing the neuronal membranes permeability to sodium ions. This reversibly stabilizes the membrane and inhibits depolarization, resulting in the failure of a propagated action potential and subsequent conduction blockade. Induces apoptosis through calpain, cytochrome oxidase etc, and shows antiviral and antiparasitic activity.