Learn More
Medchemexpress LLC HY-B0552A 500mg Medchemexpress, Dibucaine (hydrochloride) CAS:61-12-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0552A500MG
Encompass_Preferred
Medchemexpress, HY-B0552A 500mg Dibucaine (hydrochloride) CAS:61-12-1 Dibucaine hydrochloride (Cinchocaine hydrochloride) is a sodium channel inhibitor. Dibucaine hydrochloride is a potent SChE inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2596