Learn More
Medchemexpress LLC Dibucaine hydrochloride (Cinchocaine hydrochloride) | 61-12-1 | MFCD00012735 | 99.9% | 10 G
SDP
Catalog No. 5000408346
Encompass_Preferred
Dibucaine hydrochloride, also known as Cinchocaine hydrochloride, is a potent sodium channel and SChE inhibitor primarily used for research purposes. It has demonstrated the ability to reduce the degradation of BSA-gold complex in Trypanosoma cruzi reservosomes and reversibly inhibit SChE with high potency.
  • Acts as a sodium channel inhibitor
  • Identified as a potent SChE inhibitor
  • Reduced degradation of BSA-gold complex in Trypanosoma cruzi reservosomes
  • Reversibly inhibited SChE within 2 minutes
  • High potency with IC50 values of 5.3 microM (BuTch) or 3.8 microM (AcTch)
  • Competitive inhibition with BuTch (Ki of 1.3 microM)
  • Linear-mixed type inhibition with AcTch (Ki and KI of 0.66 and 2.5 microM)
  • For research use only
Catalog No. 50-004-08346 Supplier Medchemexpress LLC Supplier No. HYB0552A10G
Dibucaine hydrochloride, also known as Cinchocaine hydrochloride, is a potent sodium channel and SChE inhibitor primarily used for research purposes. It has demonstrated the ability to reduce the degradation of BSA-gold complex in Trypanosoma cruzi reservosomes and reversibly inhibit SChE with high potency.
  • Acts as a sodium channel inhibitor
  • Identified as a potent SChE inhibitor
  • Reduced degradation of BSA-gold complex in Trypanosoma cruzi reservosomes
  • Reversibly inhibited SChE within 2 minutes
  • High potency with IC50 values of 5.3 microM (BuTch) or 3.8 microM (AcTch)
  • Competitive inhibition with BuTch (Ki of 1.3 microM)
  • Linear-mixed type inhibition with AcTch (Ki and KI of 0.66 and 2.5 microM)
  • For research use only