Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl 3-hydroxybutyl phosphate | 89197-69-3 | ≥95.0% | 282.31 g/mol | C12H27O5P | 1 MG
SDP

Catalog No. 5000282119
Encompass_Preferred

Dibutyl 3-hydroxybutyl phosphate is an organophosphate metabolite formed from the metabolism of tris(n-butyl) phosphate (TNBP). It is provided as a research-use analytical reference material for studies of metabolism, environmental fate, and toxicology. Molecular formula C12H27O5P; molecular weight 282.31 g/mol.

  • Predominant metabolite of tris(n-butyl) phosphate (TNBP).
  • Intended for use as an analytical reference standard in research.
  • Applicable to metabolism, environmental fate, and toxicology studies.
  • Molecular formula C12H27O5P and molecular weight 282.31 g/mol provided.
  • Storage and handling instructions available in the certificate of analysis.

Catalog No. 50-002-82119 Supplier Medchemexpress LLC Supplier No. HYW3496281MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyl 3-hydroxybutyl phosphate is an organophosphate metabolite formed from the metabolism of tris(n-butyl) phosphate (TNBP). It is provided as a research-use analytical reference material for studies of metabolism, environmental fate, and toxicology. Molecular formula C12H27O5P; molecular weight 282.31 g/mol.

  • Predominant metabolite of tris(n-butyl) phosphate (TNBP).
  • Intended for use as an analytical reference standard in research.
  • Applicable to metabolism, environmental fate, and toxicology studies.
  • Molecular formula C12H27O5P and molecular weight 282.31 g/mol provided.
  • Storage and handling instructions available in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.