Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl adipate

Catalog No. 309494250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-949-4250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-949-4250ML Supplier Sigma Aldrich Supplier No. 309494250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyl adipate (DBA) is a plasticizer and a skin conditioning agent in cosmetics that is a mixture of a diester of butyl alcohol and adipic acid. It is soluble in water and organic solvents and does not absorb radiation in the ultraviolet region.DBA can be used as a plasticizer to produce a mixture with polyvinyl chloride (PVC), which forms soft gel type electro-active materials for soft robotic actuators and adaptive lenses. It may also be used to plasticize polyurethane gels for potential usage in biomedical applications.

Specifications

CAS 105-99-7
Density 0.962 g/mL (at 25°C)
Boiling Point 305°C
Molecular Formula C14H26O4
Refractive Index n20/D 1.436 (literature)
Linear Formula [CH2CH2CO2(CH2)3 CH3]2
Quantity 250 mL
MDL Number MFCD00009451
Molecular Weight (g/mol) 258.35
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.