Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl itaconate

Catalog No. 476196500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-619-6500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-619-6500ML Supplier Sigma Aldrich Supplier No. 476196500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyl itaconate (DBI) is an itaconate ester that can be prepared by an itaconic acid precursor.DBI is a monomer that can be copolymerized with pyridine groups to form nanocomposites for the development of bio-based elastomers. Copolymerization of DBI with acrylated epoxidized soya oil (AESO) can produce a thermoset material which finds potential applications in the modification of fatty acids.

Specifications

CAS 2155-60-4
Density 0.985 g/mL (at 25°C)
Boiling Point 284°C
Molecular Formula C13H22O4
Refractive Index n20/D 1.444 (literature)
Linear Formula CH3(CH2)3O2CCH2C(=CH2)CO2(CH2)3 CH3
MDL Number MFCD00027211
Quantity 500 mL
Synonym Di(n -butyl) itaconate; Dibutyl itaconate
Molecular Weight (g/mol) 242.31
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.