Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl Itaconate (stabilized with HQ) 97.0+%, TCI America™
SDP

Catalog No. I0201500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0201500G 500 g
I020125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0201500G Supplier TCI America Supplier No. I0201500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2155-60-4
Molecular Formula C13H22O4
Molecular Weight (g/mol) 242.315
MDL Number MFCD00027211
InChI Key OGVXYCDTRMDYOG-UHFFFAOYSA-N
Synonym Itaconic Acid Dibutyl Ester
PubChem CID 75080
IUPAC Name dibutyl 2-methylidenebutanedioate
SMILES CCCCOC(=O)CC(=C)C(=O)OCCCC

Specifications

Color Yellow
Boiling Point 190°C
Quantity 500 g
Formula Weight 242.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Itaconate (stabilized with HQ)
EINECSNumber (2)-1130
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.