Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl maleate | 105-76-0 | 99.2% | 228.28 | 100 G
SDP

Catalog No. 5000366609
Encompass_Preferred

Dibutyl maleate, the diester of Maleic Acid, is used as an intermediate in pharmaceutical synthesis. It can also enhance contact sensitization to Fluorescein isothiocyanate in mice.

  • Appearance: liquid (density: 0.988 g/cm³)
  • Color: colorless to light yellow
  • Initial source: plants Polygonaceae Rheum palmatum L.
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-66609 Supplier Medchemexpress LLC Supplier No. HYW009688100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Dibutyl maleate, the diester of Maleic Acid, is used as an intermediate in pharmaceutical synthesis. It can also enhance contact sensitization to Fluorescein isothiocyanate in mice.

  • Appearance: liquid (density: 0.988 g/cm³)
  • Color: colorless to light yellow
  • Initial source: plants Polygonaceae Rheum palmatum L.
  • Shipping: room temperature in continental US; may vary elsewhere.
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.