Learn More
Medchemexpress LLC Dibutyl maleate | 105-76-0 | 99.2% | 228.28 | 50 G
SDP
Catalog No. 5000452519
Encompass_Preferred
Dibutyl maleate, the diester of Maleic Acid, can be used as an intermediate of pharmaceutical synthesis. It can enhance contact sensitization to Fluorescein isothiocyanate in mice.
  • Can be used as an intermediate of pharmaceutical synthesis.
  • Enhances contact sensitization to Fluorescein isothiocyanate in mice.
  • For research use only.
Catalog No. 50-004-52519 Supplier Medchemexpress LLC Supplier No. HYW00968850G
Dibutyl maleate, the diester of Maleic Acid, can be used as an intermediate of pharmaceutical synthesis. It can enhance contact sensitization to Fluorescein isothiocyanate in mice.
  • Can be used as an intermediate of pharmaceutical synthesis.
  • Enhances contact sensitization to Fluorescein isothiocyanate in mice.
  • For research use only.