Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dibutyl maleate | 105-76-0 | MFCD00008544 | 99.19% | 228.28 | 1 ML
SDP

Catalog No. 5000453326
Encompass_Preferred

Dibutyl maleate is the diester of Maleic Acid. It can be used as an intermediate of pharmaceutical synthesis and is known to enhance contact sensitization to Fluorescein isothiocyanate in mice.

  • Used as an intermediate of pharmaceutical synthesis.
  • Enhances contact sensitization to fluorescein isothiocyanate in mice.
  • Molecular weight: 228.28.
  • Chemical formula: C12H20O4.
  • CAS number: 105-76-0.
  • Purity: 99.19%.
  • Appears as a colorless to light yellow liquid.
  • Soluble in DMSO (100 mg/mL).
  • Storage conditions vary based on form (pure or in solvent).

Catalog No. 50-004-53326 Supplier Medchemexpress LLC Supplier No. HYW00968810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyl maleate is the diester of Maleic Acid. It can be used as an intermediate of pharmaceutical synthesis and is known to enhance contact sensitization to Fluorescein isothiocyanate in mice.

  • Used as an intermediate of pharmaceutical synthesis.
  • Enhances contact sensitization to fluorescein isothiocyanate in mice.
  • Molecular weight: 228.28.
  • Chemical formula: C12H20O4.
  • CAS number: 105-76-0.
  • Purity: 99.19%.
  • Appears as a colorless to light yellow liquid.
  • Soluble in DMSO (100 mg/mL).
  • Storage conditions vary based on form (pure or in solvent).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.