Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dibutyl maleate

Catalog No. D47102250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47102250ML 250 mL
D471021L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47102250ML Supplier Sigma Aldrich Supplier No. D47102250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dibutyl maleate

Specifications

CAS 105-76-0
Density 0.988 g/mL (at 25°C (literature))
Boiling Point 281°C (lit.)
Molecular Formula C12H20O4
Refractive Index n20/D 1.445 (literature)
Linear Formula CH3(CH2)3 OCOCH=CHCOO(CH2)3 CH3
MDL Number MFCD00009447
Quantity 250 mL
Molecular Weight (g/mol) 228.28
Percent Purity 96%
Biological Activity Kidney
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.